missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzal chloride, 98%, Thermo Scientific Chemicals

Product Code. 11346796
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-64-2
Molecular Formula C7H5Cl2F
Molecular Weight (g/mol) 179.015
MDL Number MFCD00037112
InChI Key MVRCPAOYPLZMLB-UHFFFAOYSA-N
Synonym 3-fluorobenzal chloride, 1-dichloromethyl-3-fluorobenzene, m-fluorobenzalchloride, 3-fluorobenzalchloride, 3-fluorobenzal chlorid, acmc-1advj, 3-fluoro-1-dichloromethylbenzene, toluene,.alpha.-dichloro-m-fluoro, benzene,1-dichloromethyl-3-fluoro
PubChem CID 67875
IUPAC Name 1-(dichloromethyl)-3-fluorobenzene
SMILES C1=CC(=CC(=C1)F)C(Cl)Cl

Specifications

Boiling Point 195°C to 197°C
Refractive Index 1.5275
Quantity 5 g
UN Number UN3265
Beilstein 1936379
Formula Weight 179.02
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzal chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'