missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzylamine, 96%, Thermo Scientific Chemicals

Product Code. 11475537
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-82-3
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD00008113
InChI Key QVSVMNXRLWSNGS-UHFFFAOYSA-N
Synonym 3-fluorobenzylamine, 3-fluorophenyl methanamine, m-fluorobenzylamine, benzenemethanamine, 3-fluoro, 1-3-fluorophenyl methanamine, 3-fluoro-benzylamine, benzylamine, m-fluoro, 3-fluorophenyl methylamine, m-fluorobenzyl amine, 3-fluorobenzyl amine
PubChem CID 66853
IUPAC Name (3-fluorophenyl)methanamine
SMILES C1=CC(=CC(=C1)F)CN

Specifications

Boiling Point 186°C
Flash Point 71°C (159°F)
Odor Amine-like
Quantity 10 g
UN Number UN2735
Beilstein 1446928
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 3-Fluorobenzylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'