missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzylamine, 97%, Maybridge
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 100-82-3 |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 1.09g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C7H8FN |
Linear Formula | FC6H4CH2NH2 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'