man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
3'-Fluorobenzylspiperone maleate, Tocris Bioscience™
Print
3'-Fluorobenzylspiperone maleate, Tocris Bioscience™
D2-like receptor ligand
See promotional offers below
Description
Videos
SDS
Documents
Product Certifications
Description
Description
A very potent and selective ligand for the D2 receptor (Ki = 0.023 nM). Compared with spiperone, displays a 2.5-fold greater affinity at D2 and a 12-fold lower affinity at 5-HT2 receptors.
Videos
SDS
Documents
Documents
Product Certifications
Back to Top