missing translation for 'onlineSavingsMsg'
Learn More

(3-Fluorophenyl)methanesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10718255
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 24974-72-9
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD04117456
InChI Key QKGIPEQAFOXGSI-UHFFFAOYSA-N
Synonym 3-fluorophenyl methanesulfonyl chloride, 3-fluoro-phenyl-methanesulfonyl chloride, 3-fluorophenyl methanesulphonyl chloride, benzenemethanesulfonyl chloride, 3-fluoro, chloro 3-fluorophenyl methyl sulfone, pubchem10052, 3-fluorobenzylsulphonyl chloride, benzenemethanesulfonylchloride,3-fluoro
PubChem CID 2759107
IUPAC Name (3-fluorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)F)CS(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material (3-Fluorophenyl)methanesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'