missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine-2-carboxaldehyde, 97%, Thermo Scientific™

£20.70 - £256.00

Chemical Identifiers

CAS 31224-43-8
Molecular Formula C6H4FNO
Molecular Weight (g/mol) 125.102
MDL Number MFCD07781234
InChI Key OZIMPUNGBUYCSP-UHFFFAOYSA-N
Synonym 3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine
PubChem CID 11344017
IUPAC Name 3-fluoropyridine-2-carbaldehyde
SMILES C1=CC(=C(N=C1)C=O)F
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10021873
missing translation for 'viewDocuments'
Thermo Scientific Maybridge
CC30904CB
250 mg Amber Glass Bottle
£20.70
250mg
Please to purchase this item. Need a web account? Register with us today!
10407463
missing translation for 'viewDocuments'
Thermo Scientific Maybridge
CC30904DA
1 g Amber Glass Bottle
£32.20
1g
Please to purchase this item. Need a web account? Register with us today!
10777685
missing translation for 'viewDocuments'
Thermo Scientific Maybridge
CC30904EA
10 g Amber Glass Bottle
£256.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

31224-43-8
125.102
OZIMPUNGBUYCSP-UHFFFAOYSA-N
11344017
C1=CC(=C(N=C1)C=O)F
C6H4FNO
MFCD07781234
3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine
3-fluoropyridine-2-carbaldehyde

Specifications

31224-43-8
100.0
C6H4FNO
3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine
C1=CC(=C(N=C1)C=O)F
125.102
125.1
3-Fluoropyridine-2-carboxaldehyde
96.5
97%
MFCD07781234
OZIMPUNGBUYCSP-UHFFFAOYSA-N
3-fluoropyridine-2-carbaldehyde
11344017
97%
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Certificates