missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™
£54.60 - £164.00
Chemical Identifiers
CAS | 1073353-63-5 |
---|---|
Molecular Formula | C18H22BNO4 |
Molecular Weight (g/mol) | 327.187 |
MDL Number | MFCD09266173 |
InChI Key | CWNRULQRLNZAGF-UHFFFAOYSA-N |
Synonym | 3-furfurylaminocarbonyl benzeneboronic acid pinacol ester, n-furan-2-ylmethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-furfurylamino-1-carbonyl phenyl-boronic acid pinacol ester, 3-furfurylamino-1-carbonyl phenyl boronic acid pinacol ester, n-furan-2-yl methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb028, 3-furfurylaminocarbonyl phenylboronic acid, pinacol ester, n-furfuryl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-furan-2-ylmethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide |
PubChem CID | 46739035 |
IUPAC Name | N-(furan-2-ylmethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)NCC3=CC=CO3 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1073353-63-5 | |
327.187 | |
CWNRULQRLNZAGF-UHFFFAOYSA-N | |
46739035 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)NCC3=CC=CO3 |
C18H22BNO4 | |
MFCD09266173 | |
3-furfurylaminocarbonyl benzeneboronic acid pinacol ester, n-furan-2-ylmethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-furfurylamino-1-carbonyl phenyl-boronic acid pinacol ester, 3-furfurylamino-1-carbonyl phenyl boronic acid pinacol ester, n-furan-2-yl methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb028, 3-furfurylaminocarbonyl phenylboronic acid, pinacol ester, n-furfuryl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-furan-2-ylmethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide | |
N-(furan-2-ylmethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide |
Specifications
1073353-63-5 | |
C18H22BNO4 | |
3-furfurylaminocarbonyl benzeneboronic acid pinacol ester, n-furan-2-ylmethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-furfurylamino-1-carbonyl phenyl-boronic acid pinacol ester, 3-furfurylamino-1-carbonyl phenyl boronic acid pinacol ester, n-furan-2-yl methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb028, 3-furfurylaminocarbonyl phenylboronic acid, pinacol ester, n-furfuryl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-furan-2-ylmethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)NCC3=CC=CO3 | |
327.187 | |
327.18 | |
3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester |
122°C to 124°C | |
MFCD09266173 | |
CWNRULQRLNZAGF-UHFFFAOYSA-N | |
N-(furan-2-ylmethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide | |
46739035 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures