missing translation for 'onlineSavingsMsg'
Learn More

3-Glycidoxypropyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Product Code. 10791101
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-83-8
Molecular Formula C9H20O5Si
Molecular Weight (g/mol) 236.34
InChI Key BPSIOYPQMFLKFR-UHFFFAOYSA-N
Synonym 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
PubChem CID 17317
IUPAC Name trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane
SMILES CO[Si](CCCOCC1CO1)(OC)OC

Specifications

Density 1.0700g/mL
Boiling Point 120.0°C (2.0 mmHg)
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4280 to 1.4300
Quantity 5 g
Beilstein 17, V,3, 45
Specific Gravity 1.07
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, benzene and ether
Viscosity 4 mPa.s (20°C)
Formula Weight 236.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Glycidoxypropyltrimethoxysilane
Show More Show Less
compliance-icons
Product Identifier
  • 3-Glycidoxypropyltrimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.