Learn More
3-Hepten-2-one, 95%, Thermo Scientific Chemicals
£34.80 - £122.00
Chemical Identifiers
CAS | 1119-44-4 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.17 |
MDL Number | MFCD00015564 |
InChI Key | JHHZQADGLDKIPM-AATRIKPKSA-N |
Synonym | 3-hepten-2-one, butylideneacetone, e-hept-3-en-2-one, 3e-3-hepten-2-one, methyl pentenyl ketone, e-3-hepten-2-one, 3-hepten-2-one, e, unii-tk02t5fjdo, tk02t5fjdo, 3e-hept-3-en-2-one |
PubChem CID | 5364578 |
SMILES | CCC\C=C\C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449386
|
Thermo Scientific Alfa Aesar
A19086.09 |
10 g |
£34.80
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459386
|
Thermo Scientific Alfa Aesar
A19086.18 |
50 g |
£122.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.
Solubility
Soluble in alcohol and slightly soluble in water(3087 mg/L @ 25°C (est.))
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1119-44-4 | |
112.17 | |
JHHZQADGLDKIPM-AATRIKPKSA-N | |
5364578 |
C7H12O | |
MFCD00015564 | |
3-hepten-2-one, butylideneacetone, e-hept-3-en-2-one, 3e-3-hepten-2-one, methyl pentenyl ketone, e-3-hepten-2-one, 3-hepten-2-one, e, unii-tk02t5fjdo, tk02t5fjdo, 3e-hept-3-en-2-one | |
CCC\C=C\C(C)=O |
Specifications
1119-44-4 | |
156°C | |
C7H12O | |
MFCD00015564 | |
1739239 | |
Soluble in alcohol and slightly soluble in water(3087mg/L @ 25°C (est.)) | |
CCC\C=C\C(C)=O | |
112.17 | |
112.16 | |
3-Hepten-2-one |
0.86 | |
49°C (120°F) | |
1.445 | |
UN1224 | |
3-hepten-2-one, butylideneacetone, e-hept-3-en-2-one, 3e-3-hepten-2-one, methyl pentenyl ketone, e-3-hepten-2-one, 3-hepten-2-one, e, unii-tk02t5fjdo, tk02t5fjdo, 3e-hept-3-en-2-one | |
JHHZQADGLDKIPM-AATRIKPKSA-N | |
(E)-hept-3-en-2-one | |
5364578 | |
97% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 214-278-8
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only