Learn More
3-Hydroxy-1-methylpiperidine, 97%, Thermo Scientific Chemicals
£12.80 - £105.00
Chemical Identifiers
CAS | 3554-74-3 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.17 |
InChI Key | UKANCZCEGQDKGF-UHFFFAOYSA-N |
Synonym | 3-hydroxy-1-methylpiperidine, 1-methyl-3-piperidinol, n-methyl-3-piperidinol, 3-piperidinol, 1-methyl, 3-hydroxy-n-methylpiperidine, 1-methyl-3-hydroxypiperidine, n-methyl-3-hydroxypiperidine, n-methyl-3-hydroxy piperidine, 3r-1-methyl-3-piperidinol, pubchem6759 |
PubChem CID | 98016 |
IUPAC Name | 1-methylpiperidin-3-ol |
SMILES | CN1CCCC(C1)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3554-74-3 | |
115.17 | |
3-hydroxy-1-methylpiperidine, 1-methyl-3-piperidinol, n-methyl-3-piperidinol, 3-piperidinol, 1-methyl, 3-hydroxy-n-methylpiperidine, 1-methyl-3-hydroxypiperidine, n-methyl-3-hydroxypiperidine, n-methyl-3-hydroxy piperidine, 3r-1-methyl-3-piperidinol, pubchem6759 | |
1-methylpiperidin-3-ol |
C6H13NO | |
UKANCZCEGQDKGF-UHFFFAOYSA-N | |
98016 | |
CN1CCCC(C1)O |
Specifications
3554-74-3 | |
76.0°C to 78.0°C (11.0 mmHg) | |
96% min. (GC) | |
C6H13NO | |
0.999 | |
UKANCZCEGQDKGF-UHFFFAOYSA-N | |
1-methylpiperidin-3-ol | |
98016 | |
97% |
0.9990g/mL | |
70°C | |
Glass bottle | |
1.474 | |
3-hydroxy-1-methylpiperidine, 1-methyl-3-piperidinol, n-methyl-3-piperidinol, 3-piperidinol, 1-methyl, 3-hydroxy-n-methylpiperidine, 1-methyl-3-hydroxypiperidine, n-methyl-3-hydroxypiperidine, n-methyl-3-hydroxy piperidine, 3r-1-methyl-3-piperidinol, pubchem6759 | |
CN1CCCC(C1)O | |
115.17 | |
115.17 | |
3-Hydroxy-1-methylpiperidine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 222-609-2
- 3-Hydroxy-N-methylpiperidine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only