missing translation for 'onlineSavingsMsg'
Learn More
3-Hydroxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 11349479

3-Hydroxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11349479
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11349479

Brand: Thermo Scientific Alfa Aesar A12866.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Hydroxy-4-methoxybenzaldehyde acts as a precursor for the stereoselective synthesis of the anticancer drug (Z)-combretastatin A-4 and glycitein. It also used as an important raw material for the preparation of morphine. Further, it is involved in the preparation of Schiff- bases by reacting with furan-2-carboxylic acid hydrazide and thiophene-2-carboxylic acid hydrazide. In addition to this, it is used to prepare (Z)-2-(3-hydroxy-4-methoxybenzylidene)-1-azabicyclo[2.2.2]octan-3-one by reacting with with1-azabicyclo[2.2.2]octan-3-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-4-methoxybenzaldehyde acts as a precursor for the stereoselective synthesis of the anticancer drug (Z)-combretastatin A-4 and glycitein. It also used as an important raw material for the preparation of morphine. Further, it is involved in the preparation of Schiff- bases by reacting with furan-2-carboxylic acid hydrazide and thiophene-2-carboxylic acid hydrazide. In addition to this, it is used to prepare (Z)-2-(3-hydroxy-4-methoxybenzylidene)-1-azabicyclo[2.2.2]octan-3-one by reacting with with1-azabicyclo[2.2.2]octan-3-one.

Solubility
Soluble in acetone and methanol.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-59-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00003369
InChI Key JVTZFYYHCGSXJV-UHFFFAOYSA-N
Synonym isovanillin, isovanilline, 3-hydroxy-p-anisaldehyde, 5-formylguaiacol, benzaldehyde, 3-hydroxy-4-methoxy, iso-vanillin, 3-hydroxyanisaldehyde, isovanilin, isovanicaline, 3-hydroxy-4-methoxy-benzaldehydeShow MoreShow Less
PubChem CID 12127
IUPAC Name 3-hydroxy-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)O

Specifications

Melting Point 112°C to 116°C
Density 1.2
Boiling Point 179°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1073021
Sensitivity Air and light sensitive
Solubility Information Soluble in acetone and methanol.
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-methoxybenzaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0452100 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog NumberA12866.14,A12866.22,A12866.06
Lot NumberA0446606 Certificate TypeCertificate of Analysis Date18/11/2022 Catalog NumberA12866.14,A12866.22,A12866.06
Lot Number10239242 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA12866.14,A12866.22,A12866.06
Lot Number10239723 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA12866.14,A12866.22,A12866.06
Lot Number10223647 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12866.14,A12866.22,A12866.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Hydroxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.