missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.
Solubility
Soluble in ethanol and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under normal temperatures and pressures.
3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.
Solubility
Soluble in ethanol and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under normal temperatures and pressures.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼231°C (decomposition) |
Quantity | 1 g |
Beilstein | 2212760 |
Solubility Information | Soluble in ethanol and methanol. |
Formula Weight | 194.19 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-Hydroxy-4-methoxycinnamic acid, predominantly trans |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'