Learn More
3-Hydroxy-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
£21.10 - £163.00
Chemical Identifiers
CAS | 619-14-7 |
---|---|
Molecular Formula | C7H5NO5 |
Molecular Weight (g/mol) | 183.12 |
MDL Number | MFCD00007110 |
InChI Key | XLDLRRGZWIEEHT-UHFFFAOYSA-N |
Synonym | 3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid |
PubChem CID | 69265 |
IUPAC Name | 3-hydroxy-4-nitrobenzoic acid |
SMILES | OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499582
|
Thermo Scientific Alfa Aesar
A10724.06 |
5 g |
£21.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409592
|
Thermo Scientific Alfa Aesar
A10724.14 |
25 g |
£55.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419592
|
Thermo Scientific Alfa Aesar
A10724.22 |
100 g |
£163.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
619-14-7 | |
183.12 | |
XLDLRRGZWIEEHT-UHFFFAOYSA-N | |
69265 | |
OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O |
C7H5NO5 | |
MFCD00007110 | |
3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid | |
3-hydroxy-4-nitrobenzoic acid |
Specifications
619-14-7 | |
C7H5NO5 | |
2107564 | |
XLDLRRGZWIEEHT-UHFFFAOYSA-N | |
3-hydroxy-4-nitrobenzoic acid | |
69265 | |
98% |
232°C to 236°C | |
MFCD00007110 | |
3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid | |
OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O | |
183.12 | |
183.12 | |
3-Hydroxy-4-nitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-580-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only