Learn More
3'-Hydroxyacetophenone, 99+%, Thermo Scientific Chemicals
£15.48 - £90.75
Chemical Identifiers
CAS | 121-71-1 |
---|---|
Molecular Formula | C8H8O2 |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00002298 |
InChI Key | LUJMEECXHPYQOF-UHFFFAOYSA-N |
Synonym | 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol |
PubChem CID | 8487 |
IUPAC Name | 1-(3-hydroxyphenyl)ethanone |
SMILES | CC(=O)C1=CC(=CC=C1)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10687201
|
Thermo Scientific Acros
411910050 |
5 g | Glass bottle |
£15.48
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10526792
|
Thermo Scientific Acros
411911000 |
100 g | Glass bottle |
£90.75
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
121-71-1 | |
136.15 | |
LUJMEECXHPYQOF-UHFFFAOYSA-N | |
8487 | |
CC(=O)C1=CC(=CC=C1)O |
C8H8O2 | |
MFCD00002298 | |
3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol | |
1-(3-hydroxyphenyl)ethanone |
Specifications
121-71-1 | |
100.0 | |
Beige to Brown | |
Authentic | |
C8H8O2 | |
08, 86 | |
LUJMEECXHPYQOF-UHFFFAOYSA-N | |
1-(3-hydroxyphenyl)ethanone | |
8487 | |
99+% | |
3'-Hydroxyacetophenone, 99+% |
99.0 | |
94.0°C to 97.0°C | |
296.0°C | |
99% min. (GC) | |
MFCD00002298 | |
3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol | |
CC(=O)C1=CC(=CC=C1)O | |
136.15 | |
136.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-494-0
missing translation for 'tsca' : TSCA
RUO – Research Use Only