missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylacetic acid, 99+%, Thermo Scientific Chemicals

£51.70 - £175.00

Chemical Identifiers

CAS 621-37-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00004337
InChI Key FVMDYYGIDFPZAX-UHFFFAOYSA-N
Synonym 3-hydroxyphenylacetic acid, 2-3-hydroxyphenyl acetic acid, 3-hydroxyphenyl acetic acid, 3-hydroxybenzeneacetic acid, 3-hydroxyphenylacetate, m-hydroxyphenylacetic acid, m-hydroxyphenyl acetic acid, benzeneacetic acid, 3-hydroxy, metahydroxy phenylacetic acid, acetic acid, m-hydroxyphenyl
PubChem CID 12122
ChEBI CHEBI:17445
IUPAC Name 2-(3-hydroxyphenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(O)=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10616892
View Documents Promotion Details
Thermo Scientific Acros
121700050
5 g Glass bottle
£51.70
5g
Please to purchase this item. Need a web account? Register with us today!
10182820
View Documents Promotion Details
Thermo Scientific Acros
121700250
25 g Glass bottle
£175.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

621-37-4
152.15
FVMDYYGIDFPZAX-UHFFFAOYSA-N
12122
2-(3-hydroxyphenyl)acetic acid
C8H8O3
MFCD00004337
3-hydroxyphenylacetic acid, 2-3-hydroxyphenyl acetic acid, 3-hydroxyphenyl acetic acid, 3-hydroxybenzeneacetic acid, 3-hydroxyphenylacetate, m-hydroxyphenylacetic acid, m-hydroxyphenyl acetic acid, benzeneacetic acid, 3-hydroxy, metahydroxy phenylacetic acid, acetic acid, m-hydroxyphenyl
CHEBI:17445
OC(=O)CC1=CC=CC(O)=C1

Specifications

621-37-4
100.0
Beige-Brown to White
99% min. (HPLC)
HOC6H4CH2CO2H
10, 189
Solubility in water: soluble in water
OC(=O)CC1=CC=CC(O)=C1
152.15
CHEBI:17445
99+%
3-Hydroxyphenylacetic acid, 99+%
99.0
128.0°C to 131.0°C
Authentic
C8H8O3
MFCD00004337
3-hydroxyphenylacetic acid, 2-3-hydroxyphenyl acetic acid, 3-hydroxyphenyl acetic acid, 3-hydroxybenzeneacetic acid, 3-hydroxyphenylacetate, m-hydroxyphenylacetic acid, m-hydroxyphenyl acetic acid, benzeneacetic acid, 3-hydroxy, metahydroxy phenylacetic acid, acetic acid, m-hydroxyphenyl
FVMDYYGIDFPZAX-UHFFFAOYSA-N
2-(3-hydroxyphenyl)acetic acid
12122
152.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 210-684-4

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Hydroxyphenylacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only