missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypiperidine hydrochloride, 98+%, Thermo Scientific™

Product Code. 15452388
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Hydroxypiperidine hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64051-79-2
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.607
MDL Number MFCD00012772
InChI Key VLECDMDGMKPUSK-UHFFFAOYSA-N
Synonym 3-hydroxypiperidine hydrochloride, piperidin-3-ol hydrochloride, 3-hydroxypiperidine hcl, 3-piperidinol hydrochloride, piperidine-3-ol hydrochloride, 3-hydroxy-piperidine hydrochloride, 3-piperidinol, hydrochloride, piperidine-3-ol hcl, 3-hydroxypiperidine.hcl, 3-hydroxypiperidinehydrochloride
PubChem CID 2723962
IUPAC Name piperidin-3-ol;hydrochloride
SMILES C1CC(CNC1)O.Cl

Specifications

Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 137.61
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxypiperidine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'