missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypiperidine hydrochloride, 98+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Hydroxypiperidine hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
3-Hydroxypiperidine hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Quantity | 250 mg |
Solubility Information | Soluble in methanol. |
Formula Weight | 137.61 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-Hydroxypiperidine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'