missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.
Solubility
Soluble in methanol. Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.
Solubility
Soluble in methanol. Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Quantity | 1 g |
Beilstein | 383677 |
Solubility Information | Soluble in methanol. Slightly soluble in water. |
Formula Weight | 139.11 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 3-Hydroxypyridine-4-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'