missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxythiobenzamide, 97%, Thermo Scientific™

Product Code. 15498837
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104317-54-6
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.20
MDL Number MFCD04973331
InChI Key IDIHGRWUQGTNKS-UHFFFAOYSA-N
Synonym 3-hydroxythiobenzamide, 3-hydroxybenzene-1-carbothioamide, benzenecarbothioamide,3-hydroxy, 3-hydroxy-thiobenzamide, 3-hydroxybenzothioamide, acmc-20e42a, 3-aminothioxomethyl phenol, 3-oxidanylbenzenecarbothioamide, 3-hydroxybenzenecarbothioamide
PubChem CID 2759343
IUPAC Name 3-hydroxybenzenecarbothioamide
SMILES NC(=S)C1=CC(O)=CC=C1

Specifications

Quantity 5 g
Formula Weight 153.2
Percent Purity 97%
Chemical Name or Material 3-Hydroxythiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'