missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Iodo-L-tyrosine, 97%

Catalog Number p-3776699
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
70-78-0
C9H10INO3
307.09
MFCD00002608
UQTZMGFTRHFAAM-JLDDOWRYNA-N
3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
439744
CHEBI:27847
(2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O
This item is not returnable. View return policy
70-78-0
C9H10INO3
307.09
MFCD00002608
UQTZMGFTRHFAAM-JLDDOWRYNA-N
3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
439744
CHEBI:27847
(2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 196.0°C to 198.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Heavy Metals (as Pb) 20ppm max.
Assay Percent Range 97%
Infrared Spectrum Authentic
Merck Index 15, 5090
Packaging Glass Bottle
Solubility Information Solubility in water: .. Other solubilities: soluble in 15 parts boiling water
Specific Rotation − 3.18
Formula Weight 307.08
Percent Purity 97%
Water 1% max. (K.F.)
Quantity 1 g
Physical Form Powder
Chemical Name or Material 3-Iodo-L-tyrosine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'