missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-L-tyrosine, 98%, Thermo Scientific Chemicals
£30.30 - £112.00
Chemical Identifiers
CAS | 70-78-0 |
---|---|
Molecular Formula | C9H10INO3 |
Molecular Weight (g/mol) | 307.09 |
MDL Number | MFCD00002608 |
InChI Key | UQTZMGFTRHFAAM-JLDDOWRYNA-N |
Synonym | 3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine |
PubChem CID | 439744 |
ChEBI | CHEBI:27847 |
IUPAC Name | (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid |
SMILES | N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15410679
|
Thermo Scientific Alfa Aesar
H63789.03 |
1 g |
£30.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420679
|
Thermo Scientific Alfa Aesar
H63789.06 |
5 g |
£112.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.
Solubility
Soluble in dilute aqeous acid and dilute aqeous base.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
Chemical Identifiers
70-78-0 | |
307.09 | |
UQTZMGFTRHFAAM-JLDDOWRYNA-N | |
439744 | |
(2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid |
C9H10INO3 | |
MFCD00002608 | |
3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine | |
CHEBI:27847 | |
N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O |
Specifications
210°C (decomposition) | |
White | |
MFCD00002608 | |
3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine | |
UQTZMGFTRHFAAM-JLDDOWRYNA-N | |
(2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid | |
439744 | |
307.09 | |
Light sensitive | |
3-Iodo-L-tyrosine |
70-78-0 | |
C9H10INO3 | |
2941266 | |
Soluble in dilute aqeous acid and dilute aqeous base. | |
N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O | |
307.09 | |
CHEBI:27847 | |
98% | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 200-744-8
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only