missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15467997
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221037-98-5
Molecular Formula C6H6BIO2
Molecular Weight (g/mol) 247.826
MDL Number MFCD01319015
InChI Key REEUXWXIMNEIIN-UHFFFAOYSA-N
Synonym 3-iodophenyl boronic acid, boronic acid, 3-iodophenyl, 3-iodobenzeneboronic acid, pentanedioic acid 1-1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl ester, pubchem6224, acmc-1ckkp, 3-iodophenylboranic acid, amtb078, 3-iodanylphenyl boronic acid, boronic acid,b-3-iodophenyl
PubChem CID 2734362
IUPAC Name (3-iodophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)I)(O)O

Specifications

Quantity 25 g
Sensitivity Light sensitive
Formula Weight 247.83
Percent Purity 97%
Chemical Name or Material 3-Iodobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'