missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzyl bromide, 97%, Thermo Scientific Chemicals

Product Code. 10702095
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49617-83-6
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.93
MDL Number MFCD00019016
InChI Key BACZSVQZBSCWIG-UHFFFAOYSA-N
Synonym 3-iodobenzyl bromide, 1-bromomethyl-3-iodobenzene, alpha-bromo-3-iodotoluene, 3-iodobenzylbromide, m-iodobenzyl bromide, benzene, 1-bromomethyl-3-iodo, m-iodobenzylbromide, 3-iodo-benzylbromide, 3-iodo-benzyl bromide, meta-iodobenzyl bromide
PubChem CID 2759361
IUPAC Name 1-(bromomethyl)-3-iodobenzene
SMILES C1=CC(=CC(=C1)I)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 47°C to 50°C
Color Brown or Orange to Red
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4I
Quantity 25 g
Formula Weight 296.93
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3-Iodobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'