missing translation for 'onlineSavingsMsg'
Learn More

(3-Isocyanatopropyl)triethoxysilane, 95%, Thermo Scientific Chemicals

£37.49 - £104.52

Chemical Identifiers

CAS 24801-88-5
Molecular Formula C10H21NO4Si
Molecular Weight (g/mol) 247.37
MDL Number MFCD00051459
InChI Key FRGPKMWIYVTFIQ-UHFFFAOYSA-N
Synonym 3-isocyanatopropyltriethoxysilane, triethoxy 3-isocyanatopropyl silane, 3-isocyanatopropyl triethoxysilane, isocyanatopropyltriethoxysilane, 3-triethoxysilyl propyl isocyanate, silane, triethoxy 3-isocyanatopropyl, unii-9br6002p6e, isocyanic acid, 3-triethoxysilyl propylester, isocyanic acid, 3-triethoxysilyl propyl ester, icptesShow MoreShow Less
PubChem CID 90613
IUPAC Name triethoxy(3-isocyanatopropyl)silane
SMILES CCO[Si](CCCN=C=O)(OCC)OCC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10688414
SDS View Documents
Thermo Scientific Acros
391900250
25 g Glass bottle
£37.49
25g
Please to purchase this item. Need a web account? Register with us today!
10279093
SDS View Documents
Thermo Scientific Acros
391901000
100 g Glass bottle
£104.52
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 246-467-6

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(3-Isocyanatopropyl)triethoxysilane, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.