missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropylbenzeneboronic acid, 99%, Thermo Scientific Chemicals

Product Code. 11348316
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216019-28-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD01074634
InChI Key QSWLFBMVIGQONC-UHFFFAOYSA-N
Synonym 3-isopropylphenylboronic acid, 3-isopropylbenzeneboronic acid, 3-cumylboronic acid, 3-isopropylphenyl boronic acid, 3-isopropylphebylboronic acid, 3-propan-2-yl phenyl boranediol, boronic acid, 3-1-methylethyl phenyl, boronic acid, b-3-propylphenyl, 3-propan-2-yl phenyl boronic acid, 3-propan-2-ylphenyl boronic acid
PubChem CID 2773478
IUPAC Name (3-propan-2-ylphenyl)boronic acid
SMILES CC(C)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point ∼75°C to 80°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 3090001
Formula Weight 164.01
Percent Purity 99%
Chemical Name or Material 3-Isopropylbenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'