missing translation for 'onlineSavingsMsg'
Learn More
3-Methoxy-5-methylphenol, 97%, Thermo Scientific Chemicals Product Code.: 11431780

3-Methoxy-5-methylphenol, 97%, Thermo Scientific Chemicals

Product Code. 11431780
5 g
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11431780

Brand: Thermo Scientific Alfa Aesar H27516.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3209-13-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00059261
InChI Key NOTCZLKDULMKBR-UHFFFAOYSA-N
Synonym 3-hydroxy-5-methoxytoluene, phenol, 3-methoxy-5-methyl, 5-methoxy-m-cresol, orcinol monomethyl ether, o-methylorcinol, 3-methoxy-5-methyl-phenol, unii-uya9w3l847, 5-methoxy-3-methylphenol, orcinyl ter, 3-methoxy-5-methylphenol
PubChem CID 76674
IUPAC Name 3-methoxy-5-methylphenol
SMILES CC1=CC(=CC(=C1)OC)O

Specifications

Melting Point 60°C to 66°C
Quantity 5 g
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-methylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methoxy-5-methylphenol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.