missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 15465147
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53985-48-1
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD09025409
InChI Key VAQSHRZOMRFBCX-UHFFFAOYSA-N
Synonym 3-methoxy-5-trifluoromethyl benzoic acid, 3-methoxy-5-trifluoromethyl-benzoic acid, benzoic acid, 3-methoxy-5-trifluoromethyl, 3-carboxy-5-methoxybenzotrifluoride, 3-carboxy-5-trifluoromethyl anisole, 3methoxy-5-trifluoromethylbenzoic acid, 3-methoxy-5-trifluoromethyl benzoicacid, 5-methoxy-3-trifluoromethyl benzoic acid, 3-trifluoromethyl-5-methoxybenzoic acid
PubChem CID 21544467
IUPAC Name 3-methoxy-5-(trifluoromethyl)benzoic acid
SMILES COC1=CC(=CC(=C1)C(F)(F)F)C(=O)O

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'