missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-N,N-dimethylaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
N,N-Dimethyl-m-anisidine is a phenyl amide with antineoplastic and antibacterial properties. It can be used to produce C20H19NO4. It will need reagent tributyl orthoformate and solvent propan-2-ol with reaction time of 90 min.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethyl-m-anisidine is a phenyl amide with antineoplastic and antibacterial properties. It can be used to produce C20H19NO4. It will need reagent tributyl orthoformate and solvent propan-2-ol with reaction time of 90 min.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
N,N-Dimethyl-m-anisidine is a phenyl amide with antineoplastic and antibacterial properties. It can be used to produce C20H19NO4. It will need reagent tributyl orthoformate and solvent propan-2-ol with reaction time of 90 min.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Density | 1.031 |
Boiling Point | 113°C to 114°C (6 mmHg) |
Refractive Index | 1.559 |
Quantity | 10 g |
UN Number | UN2810 |
Beilstein | 1423591 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 151.21 |
Percent Purity | 98% |
Chemical Name or Material | 3-Methoxy-N,N-dimethylaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'