Learn More
3-Methoxybenzyl chloride, 98+%, Thermo Scientific Chemicals
£25.30 - £606.00
Chemical Identifiers
CAS | 824-98-6 |
---|---|
Molecular Formula | C8H9ClO |
Molecular Weight (g/mol) | 156.61 |
MDL Number | MFCD00000907 |
InChI Key | VGISFWWEOGVMED-UHFFFAOYSA-N |
Synonym | 3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy |
PubChem CID | 69994 |
IUPAC Name | 1-(chloromethyl)-3-methoxybenzene |
SMILES | COC1=CC=CC(CCl)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379469
|
Thermo Scientific Alfa Aesar
A12630.06 |
5 g |
£25.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389469
|
Thermo Scientific Alfa Aesar
A12630.14 |
25 g |
£70.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470463
|
Thermo Scientific Alfa Aesar
A12630.18 |
50 g |
£116.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480463
|
Thermo Scientific Alfa Aesar
A12630.30 |
250 g |
£606.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
824-98-6 | |
156.61 | |
VGISFWWEOGVMED-UHFFFAOYSA-N | |
69994 | |
COC1=CC=CC(CCl)=C1 |
C8H9ClO | |
MFCD00000907 | |
3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy | |
1-(chloromethyl)-3-methoxybenzene |
Specifications
824-98-6 | |
101°C (213°F) | |
MFCD00000907 | |
636684 | |
VGISFWWEOGVMED-UHFFFAOYSA-N | |
1-(chloromethyl)-3-methoxybenzene | |
69994 | |
≥98% |
122°C to 123°C (12 mmHg) | |
C8H9ClO | |
UN1760 | |
3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy | |
COC1=CC=CC(CCl)=C1 | |
156.61 | |
156.61 | |
3-Methoxybenzyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Recommended Storage : 2° to 8°C
RUO – Research Use Only