missing translation for 'onlineSavingsMsg'
Learn More

3-(Methoxycarbonyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11349485
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41221-47-0
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00037077
InChI Key WBGWGERFPSYHDT-UHFFFAOYSA-N
Synonym 3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate
PubChem CID 2733366
IUPAC Name methyl 3-isocyanatobenzoate
SMILES COC(=O)C1=CC=CC(=C1)N=C=O

Specifications

Melting Point 35°C to 38°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2811
Sensitivity Moisture sensitive
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 3-(Methoxycarbonyl)phenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(Methoxycarbonyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.