missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylglyoxal hydrate, 97%, dry wt. basis, Thermo Scientific Chemicals

Product Code. 11389126
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32025-65-3
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD08533286
InChI Key SIHYQEYAJMDKQH-UHFFFAOYSA-N
Synonym 3-methoxyphenylglyoxal hydrate, 2-3-methoxyphenyl-2-oxoacetaldehyde hydrate, acmc-1c24s, bestipharma 555-843, 3-methoxyphenylglyoxal monohydrate, ethanone,2,2-dihydroxy-1-3-methylphenyl, 3-methoxyphenyl oxo acetaldehyde-water 1/1, 2-3-methoxyphenyl-2-oxidanylidene-ethanal hydrate
PubChem CID 21155397
IUPAC Name 2-(3-methoxyphenyl)-2-oxoacetaldehyde;hydrate
SMILES COC1=CC=CC(=C1)C(=O)C=O

Specifications

Melting Point 94°C to 98°C
Boiling Point 95°C to 98°C (0.05 mmHg)
Quantity 5 g
Beilstein 1935225
Formula Weight 164.16 (Anhydrous)
Percent Purity 97%
Assay (dry wt.)
Chemical Name or Material 3-Methoxyphenylglyoxal hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'