missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylisothiocyanate, ≥97%, Thermo Scientific™

Product Code. 10483014
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 3125-64-2
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00040335
InChI Key WHBYCPUKGYEYFU-UHFFFAOYSA-N
Synonym 3-methoxyphenyl isothiocyanate, 3-methoxyphenylisothiocyanate, 1-isothiocyanato-3-methoxy-benzene, 3-isothiocyanatoanisole, acmc-1cldj, m-methoxyphenyl isothiocyanate, 3-methoxy phenyl isothiocyanate, 3-methoxy-phenyl isothiocyanate, #
PubChem CID 137832
IUPAC Name 1-isothiocyanato-3-methoxybenzene
SMILES COC1=CC=CC(=C1)N=C=S

Specifications

CAS Min % 97.0
Density 1.1790g/mL
Assay Percent Range 97+%
Linear Formula CH3OC6H4NCS
Quantity 25 g
Formula Weight 165.22
Percent Purity 97%
Chemical Name or Material 3-Methoxyphenylisothiocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.