missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropylamine solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

£46.55 - £96.60

Chemical Identifiers

CAS 5332-73-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00014831
InChI Key FAXDZWQIWUSWJH-UHFFFAOYSA-N
PubChem CID 1672
IUPAC Name 3-methoxypropan-1-amine
SMILES COCCCN
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12902075
View Documents Promotion Details
Fisher Chemical
A 001 100
100 mL Bottle
£46.55
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12912075
View Documents Promotion Details
Fisher Chemical
A 001 250
250 mL Bottle
£66.35
250mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12922075
View Documents Promotion Details
Fisher Chemical
A 001 500
500 mL Bottle
£96.60
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

5332-73-0
89.14
FAXDZWQIWUSWJH-UHFFFAOYSA-N
3-methoxypropan-1-amine
C4H11NO
MFCD00014831
1672
COCCCN

Specifications

1g/L
Single-component Standard Solution
MFCD00014831
COCCCN
89.14
H2O
3-Methoxypropylamine
5332-73-0
C4H11NO
FAXDZWQIWUSWJH-UHFFFAOYSA-N
3-methoxypropan-1-amine
1672
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months. (12 Months after opening)

SDS
Documents

Documents

Product Certifications