missing translation for 'onlineSavingsMsg'
Learn More
3-Methyl-1,2-cyclopentanediol, mixture of diastereomers, 95%, Thermo Scientific™ Product Code.: 11464397

3-Methyl-1,2-cyclopentanediol, mixture of diastereomers, 95%, Thermo Scientific™

Product Code. 11464397
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11464397

Brand: Thermo Scientific Alfa Aesar B20440.03

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27583-37-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00019285
InChI Key KANFKJUPLALTDB-UHFFFAOYNA-N
Synonym 3-methyl-1,2-cyclopentanediol, 1,2-cyclopentanediol, 3-methyl, 3-methylcyclopentan-1,2-diol, 3-methyl-1,2-cyclopentanediol, mixture of diastereomers, 3-methylcyclopentanediol, acmc-20an1b, 3-methyl-1,2-cyclopentanediol #, 3-methyl-1,2-cyclopentanediol mixture of isomers
PubChem CID 33945
IUPAC Name 3-methylcyclopentane-1,2-diol
SMILES CC1CCC(O)C1O

Specifications

Quantity 1 g
Formula Weight 116.16
Percent Purity 95%
Chemical Name or Material 3-Methyl-1,2-cyclopentanediol, mixture of diastereomers

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH5692A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberF2170B Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberE4538C Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-1,2-cyclopentanediol, mixture of diastereomers, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.