missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1-Butanol, 98%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | -117°C |
Color | Colorless |
Density | 0.809g/mL at 25°C |
Boiling Point | 131°C to 132°C |
Explosion Limit | 1.2 to 9% (100°F) |
Flash Point | 43°C (109.4°F) |
Ignition Point | 644°F |
Packaging | Glass bottle |
Linear Formula | (CH3)2CHCH2CH2OH |
Quantity | 2.5 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'