missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-phospholene 1-oxide, 94%, Thermo Scientific Chemicals

Product Code. 11469373
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

Solubility
Soluble in polar solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-61-9
Molecular Formula C11H13OP
Molecular Weight (g/mol) 192.198
MDL Number MFCD00014518
InChI Key YMKWWHFRGALXLE-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-2-phospholene 1-oxide, 3-methyl-1-phenyl-2-phospholene-1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole 1-oxide, 1h-phosphole, 2,3-dihydro-4-methyl-1-phenyl-, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1h-phosphole 1-oxide, 2-phospholene, 3-methyl-1-phenyl-, 1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1$l^ 5-phosphole 1-oxide, 3-methyl-1-phenyl-4,5-dihydro-1??-phosphol-1-one, 2-phospholene, 1-oxide
PubChem CID 69722
IUPAC Name 4-methyl-1-phenyl-2,3-dihydro-1$l^{5}-phosphole 1-oxide
SMILES CC1=CP(=O)(CC1)C2=CC=CC=C2

Specifications

Melting Point 60°C to 65°C
Boiling Point 173°C to 174°C (0.7 mmHg)
Odor Unpleasant
Quantity 5 g
Beilstein 609223
Sensitivity Hygroscopic
Solubility Information Soluble in polar solvents.
Formula Weight 192.19
Percent Purity 94%
Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.