missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-pyrazolin-5-one, 98+%, Thermo Scientific Chemicals

Product Code. 11499083
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

Solubility
Soluble in water(3.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-25-8
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00003138
InChI Key QELUYTUMUWHWMC-UHFFFAOYSA-N
Synonym edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
PubChem CID 4021
ChEBI CHEBI:31530
IUPAC Name 5-methyl-2-phenyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1)C2=CC=CC=C2

Specifications

Melting Point 127°C to 131°C
Density 1.12
Boiling Point 191°C (17 mmHg)
Odor Odorless
Quantity 100 g
Beilstein 609575
Merck Index 14,6713
Solubility Information Soluble in water(3.3g/L).
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-1-phenyl-2-pyrazolin-5-one
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-1-phenyl-2-pyrazolin-5-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 3-Methyl-1-phenyl-2-pyrazolin-5-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'