missing translation for 'onlineSavingsMsg'
Learn More
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%, Thermo Scientific Chemicals Product Code.: 11437787

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%, Thermo Scientific Chemicals

Product Code. 11437787
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11437787

Brand: Thermo Scientific Alfa Aesar A16317.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149022-15-1
Molecular Formula C8H10ClN3S
Molecular Weight (g/mol) 215.70
MDL Number MFCD00149370
InChI Key OEZPVSPULCMUQB-UHFFFAOYSA-N
Synonym unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
PubChem CID 9575839
IUPAC Name (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride
SMILES Cl.CN1C(SC2=CC=CC=C12)=NN

Specifications

Melting Point ∼275°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3571041
Solubility Information Soluble in water,dimethyl sulfoxide and methanol.
Formula Weight 215.71 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate
compliance-icons
Product Identifier
  • 3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : 3-Methyl-2-benzothiazolinone hydrazone hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.