Learn More
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%, Thermo Scientific Chemicals
Beschreibung
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.
Solubility
Soluble in water, dimethyl sulfoxide and methanol.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
Melting Point | ∼275°C (decomposition) |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 3571041 |
Solubility Information | Soluble in water,dimethyl sulfoxide and methanol. |
Formula Weight | 215.71 (Anhydrous) |
Percent Purity | 97% |
Chemical Name or Material | 3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate |
RUO – Research Use Only