missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-2-cyclohexen-1-one, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
Description
3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
Specifications
Density | 0.971 |
Boiling Point | 199°C to 200°C |
Flash Point | 68°C (154°F) |
Odor | Pleasant |
Refractive Index | 1.494 |
Quantity | 10 g |
Beilstein | 1560601 |
Solubility Information | Soluble in alcohol. Insoluble in water. |
Formula Weight | 110.15 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'