Learn More
3-Methyl-2-cyclohexen-1-one, 98%, Thermo Scientific Chemicals
£39.70 - £225.00
Chemical Identifiers
CAS | 1193-18-6 |
---|---|
Molecular Formula | C7H10O |
Molecular Weight (g/mol) | 110.16 |
MDL Number | MFCD00001581 |
InChI Key | IITQJMYAYSNIMI-UHFFFAOYSA-N |
Synonym | 3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699 |
PubChem CID | 14511 |
IUPAC Name | 3-methylcyclohex-2-en-1-one |
SMILES | CC1=CC(=O)CCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349469
|
Thermo Scientific Alfa Aesar
A15704.09 |
10 g |
£39.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487667
|
Thermo Scientific Alfa Aesar
A15704.14 |
25 g |
£68.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359469
|
Thermo Scientific Alfa Aesar
A15704.18 |
50 g |
£121.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11497667
|
Thermo Scientific Alfa Aesar
A15704.22 |
100 g |
£225.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.
Chemical Identifiers
1193-18-6 | |
110.16 | |
IITQJMYAYSNIMI-UHFFFAOYSA-N | |
14511 | |
CC1=CC(=O)CCC1 |
C7H10O | |
MFCD00001581 | |
3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699 | |
3-methylcyclohex-2-en-1-one |
Specifications
1193-18-6 | |
199°C to 200°C | |
Pleasant | |
1.494 | |
1560601 | |
Soluble in alcohol. Insoluble in water. | |
CC1=CC(=O)CCC1 | |
110.16 | |
110.15 | |
3-Methyl-2-cyclohexen-1-one |
0.971 | |
68°C (154°F) | |
C7H10O | |
MFCD00001581 | |
3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699 | |
IITQJMYAYSNIMI-UHFFFAOYSA-N | |
3-methylcyclohex-2-en-1-one | |
14511 | |
98% |
Safety and Handling
GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
H227-H302
missing translation for 'einecsNumber' : 214-769-7
missing translation for 'rtecsNumber' : GW7340000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only