missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclohexen-1-one, 98%, Thermo Scientific Chemicals

£17.80 - £114.00

Chemical Identifiers

CAS 1193-18-6
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00001581
InChI Key IITQJMYAYSNIMI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
PubChem CID 14511
IUPAC Name 3-methylcyclohex-2-en-1-one
SMILES CC1=CC(=O)CCC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10043303
View Documents Promotion Details
Thermo Scientific Acros
126660010
1 g Glass bottle
£17.80
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10469050
View Documents Promotion Details
Thermo Scientific Acros
126660100
10 g Glass bottle
£37.40
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10350114
View Documents Promotion Details
Thermo Scientific Acros
126660500
50 g Glass bottle
£114.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1193-18-6
110.16
IITQJMYAYSNIMI-UHFFFAOYSA-N
14511
CC1=CC(=O)CCC1
C7H10O
MFCD00001581
3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
3-methylcyclohex-2-en-1-one

Specifications

1193-18-6
100.0
Brown to Yellow
199.0°C to 200.0°C
Authentic
C7H10O
1.4935 to 1.4955
07, 54
3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
IITQJMYAYSNIMI-UHFFFAOYSA-N
3-methylcyclohex-2-en-1-one
14511
98%
3-Methyl-2-cyclohexen-1-one
97.5
-21°C
0.9700g/mL
74°C
97.5% min. (GC)
CH3C6H7(=O)
MFCD00001581
0.97
Solubility in water: insoluble
CC1=CC(=O)CCC1
110.16
110.16
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 214-769-7

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only