missing translation for 'onlineSavingsMsg'
Learn More

(3-Methyl-2-furyl)methylamine, 97%, Thermo Scientific™

£80.55 - £219.00

Chemical Identifiers

CAS 388072-09-1
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.144
MDL Number MFCD09811371
InChI Key CTGIVQJZUKJDNH-UHFFFAOYSA-N
Synonym 3-methylfuran-2-yl methanamine, 3-methyl-2-furyl methylamine, 2-aminomethyl-3-methylfuran, 2-furanmethanamine,3-methyl, 2-furanmethanamine, 3-methyl, 1-3-methylfuran-2-yl methanamine, 3-methylfurfurylamine, 3-methylfur-2-yl methylamine
PubChem CID 18187367
IUPAC Name (3-methylfuran-2-yl)methanamine
SMILES CC1=C(OC=C1)CN
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10535554
View Documents Promotion Details
Thermo Scientific Maybridge
CC55513CB
250 mg Amber Glass Bottle
£80.55
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10738635
View Documents Promotion Details
Thermo Scientific Maybridge
CC55513DA
1 g Amber Glass Bottle
£219.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

388072-09-1
111.144
CTGIVQJZUKJDNH-UHFFFAOYSA-N
18187367
CC1=C(OC=C1)CN
C6H9NO
MFCD09811371
3-methylfuran-2-yl methanamine, 3-methyl-2-furyl methylamine, 2-aminomethyl-3-methylfuran, 2-furanmethanamine,3-methyl, 2-furanmethanamine, 3-methyl, 1-3-methylfuran-2-yl methanamine, 3-methylfurfurylamine, 3-methylfur-2-yl methylamine
(3-methylfuran-2-yl)methanamine

Specifications

388072-09-1
100.0
C6H9NO
3-methylfuran-2-yl methanamine, 3-methyl-2-furyl methylamine, 2-aminomethyl-3-methylfuran, 2-furanmethanamine,3-methyl, 2-furanmethanamine, 3-methyl, 1-3-methylfuran-2-yl methanamine, 3-methylfurfurylamine, 3-methylfur-2-yl methylamine
CC1=C(OC=C1)CN
111.144
111.14
(3-Methyl-2-furyl)methylamine
96.5
97%
MFCD09811371
CTGIVQJZUKJDNH-UHFFFAOYSA-N
(3-methylfuran-2-yl)methanamine
18187367
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a

Danger

SDS
Documents

Documents

Product Certifications