missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pyridone, 97+%, Thermo Scientific™

Product Code. 11450613
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Methyl-2-pyridone, is used as a reactant with chloro-methoxy-methane to produce 1-methoxymethyl-3-methyl-1H-pyridin-2-one. This reaction will need reagent solvent CH2Cl2. The reaction time is 24 hours with ambient temperature

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-56-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00039704
InChI Key MVKDNXIKAWKCCS-UHFFFAOYSA-N
Synonym 3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone
PubChem CID 70481
IUPAC Name 3-methyl-1H-pyridin-2-one
SMILES CC1=CC=CNC1=O

Specifications

Melting Point 137°C to 142°C
Quantity 5 g
Beilstein 1422044
Solubility Information Soluble in methanol.
Formula Weight 109.13
Percent Purity ≥97%
Chemical Name or Material 3-Methyl-2-pyridone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'