missing translation for 'onlineSavingsMsg'
Learn More
3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane, 98%, Thermo Scientific Chemicals Product Code.: 15418646

3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane, 98%, Thermo Scientific Chemicals

Product Code. 15418646
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15418646

Brand: Thermo Scientific Alfa Aesar H64011.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99314-44-0
Molecular Formula C12H16O4S
Molecular Weight (g/mol) 256.316
MDL Number MFCD08275394
InChI Key PZOQQSOZRVZCMC-UHFFFAOYSA-N
Synonym 3-methyloxetan-3-yl methyl 4-methylbenzenesulfonate, 3-methyl-3-oxetanyl methyl 4-methylbenzenesulfonate, 3-methyloxetan-3-yl methyl p-tosylate, 3-methyl-3-toluenesulfonyloxymethyl oxetane, 3-methyloxetan-3-yl methyl 4-methylbenzene-1-sulfonate, upcmld00wcrh3-139, 3-tosyloxymethyl-3-methyloxetane, 3-methyl-3-oxetanyl methyl p-tosylate, methyloxetanylmethylmethylbenzenesulfonate, 3-methyloxetan-3-yl methyl 4-toluenesulfonate
PubChem CID 5461213
IUPAC Name (3-methyloxetan-3-yl)methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2(COC2)C

Specifications

Melting Point 50°C to 51°C
Color White
Quantity 5 g
Beilstein 4441465
Formula Weight 256.32
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Methyl-3-(p-toluenesulfonyloxymethyl)oxetane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.