missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-trimethylsiloxy-1-butyne, 97%, Thermo Scientific Chemicals

Product Code. 11364968
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17869-77-1
Molecular Formula C8H16OSi
Molecular Weight (g/mol) 156.3
MDL Number MFCD00053867
InChI Key JNRUXZIXAXHXTN-UHFFFAOYSA-N
Synonym 3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne
PubChem CID 87344
IUPAC Name trimethyl(2-methylbut-3-yn-2-yloxy)silane
SMILES CC(C)(C#C)O[Si](C)(C)C

Specifications

Boiling Point 52°C (75 mmHg)
Odor Characteristic
Refractive Index 1.4052
Quantity 10 g
UN Number UN1993
Formula Weight 156.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-trimethylsiloxy-1-butyne

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'