Learn More
3-Methyl-3-trimethylsiloxy-1-butyne, 97%, Thermo Scientific Chemicals
£27.00 - £413.00
Chemical Identifiers
CAS | 17869-77-1 |
---|---|
Molecular Formula | C8H16OSi |
Molecular Weight (g/mol) | 156.3 |
MDL Number | MFCD00053867 |
InChI Key | JNRUXZIXAXHXTN-UHFFFAOYSA-N |
Synonym | 3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne |
PubChem CID | 87344 |
IUPAC Name | trimethyl(2-methylbut-3-yn-2-yloxy)silane |
SMILES | CC(C)(C#C)O[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374968
|
Thermo Scientific Alfa Aesar
043922.04 |
2 g |
£27.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364968
|
Thermo Scientific Alfa Aesar
043922.09 |
10 g |
£80.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354968
|
Thermo Scientific Alfa Aesar
043922.18 |
50 g |
£413.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17869-77-1 | |
156.3 | |
JNRUXZIXAXHXTN-UHFFFAOYSA-N | |
87344 | |
CC(C)(C#C)O[Si](C)(C)C |
C8H16OSi | |
MFCD00053867 | |
3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne | |
trimethyl(2-methylbut-3-yn-2-yloxy)silane |
Specifications
17869-77-1 | |
Characteristic | |
1.4052 | |
UN1993 | |
JNRUXZIXAXHXTN-UHFFFAOYSA-N | |
trimethyl(2-methylbut-3-yn-2-yloxy)silane | |
87344 | |
97% | |
3-Methyl-3-trimethylsiloxy-1-butyne |
52°C (75 mmHg) | |
C8H16OSi | |
MFCD00053867 | |
3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne | |
CC(C)(C#C)O[Si](C)(C)C | |
156.3 | |
156.3 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RUO – Research Use Only