missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzamide, 97%, Thermo Scientific™

Product Code. 11480568
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Methyl-4-nitrobenzamide is used as synthetic organic intermediates.

Solubility
Soluble in alcohol, slightly soluble in hot water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99584-85-7
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.16
MDL Number MFCD00031468
InChI Key BCAAGQMIPKPDDX-UHFFFAOYSA-N
Synonym 3-methyl-4-nitro-benzamide, 3-methyl-4-nitro benzamide, #, benzamide, 3-methyl-4-nitro, 3-methyl-4-nitrobenzoic acid amide
PubChem CID 595900
IUPAC Name 3-methyl-4-nitrobenzamide
SMILES CC1=CC(=CC=C1[N+]([O-])=O)C(N)=O

Specifications

Melting Point 158°C to 160°C
Quantity 1 g
Beilstein 2264048
Solubility Information Soluble in alcohol,slightly soluble in hot water.
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'