missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-4-nitrophenol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Descripción
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.
Solubility
Soluble in water(1.34g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.
Solubility
Soluble in water(1.34g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
Melting Point | 126°C to 130°C |
Odor | Phenol-like |
Quantity | 100 g |
Beilstein | 1868105 |
Solubility Information | Soluble in water(1.34g/L). |
Formula Weight | 153.14 |
Percent Purity | 98% |
Chemical Name or Material | 3-Methyl-4-nitrophenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'