missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15498865
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1240257-48-0
Molecular Formula C8H8BrF5S
Molecular Weight (g/mol) 311.108
MDL Number MFCD16652481
InChI Key ZRHPUHTTXXCZFI-UHFFFAOYSA-N
Synonym 3-methyl-5-pentafluorosulfur benzyl bromide, 3-methyl-5-pentafluorothio benzyl bromide, 1-bromomethyl-3-methyl-5-pentafluoro-??-sulfanyl benzene
PubChem CID 66523600
IUPAC Name [3-(bromomethyl)-5-methylphenyl]-pentafluoro-$l^{6}-sulfane
SMILES CC1=CC(=CC(=C1)S(F)(F)(F)(F)F)CBr

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 311.11
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(pentafluorothio)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'